Polyvinyl alcohol, M.W. ∼ 146,000-186,000 92-94%, hydrolysed

Supplier: Thermo Scientific Chemicals

18316-0010 183160025 183161000
TS18316-0010EA 132 USD
TS18316-0010 TS183160025 TS183161000
Polyvinyl alcohol, M.W. ∼ 146,000-186,000 92-94%, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White to cream Granular powder
Infrared spectrum Conforms
Viscosity 53 to 63 cP (4 % in water)
Degree of hydrolysis 92.0 to 94.0 mol %
Ash ≤0.6 % (as Na2O)
pH 4.5 to 6.5 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR