Acrylamide polymer, M.W. M.W. ~ 5 -6.000.000, granules, non-ionic

Supplier: Thermo Scientific Chemicals

17804-2500 178040050 178045000
TS17804-2500EA 287 USD
TS17804-2500 TS178040050 TS178045000
Acrylamide polymer, M.W. M.W. ~ 5 -6.000.000, granules, non-ionic
Acrylamide polymer

Appearance: White to light yellow Granules

Formula: (CH₂:CHCONH₂)n
Density: 1.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084392
CAS Number: 9003-05-8

Order Now

Specification Test Results

Appearance White to light yellow Granules
Infrared spectrum Conforms
Loss on drying ≤13 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR