Benzonitrile 99+% for spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
16767-0500
167670010
167672500
TS16767-0500EA
41
USD
TS16767-0500
TS167670010
TS167672500
Benzonitrile 99+% for spectroscopy
Benzonitrile
Formula:
C₇H₅N Boiling Pt: 188…191 °C (760 mmHg) Melting Pt: –13 °C Density: 1.01 g/cm³ Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001770 CAS Number: 100-47-0 EINECS: 202-855-7 UN: 2224 ADR: 6.1,II Merck Index: 13,01098 |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.03 % |
GC | ≥99.0 % |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...