Trifluoroacetic acid-[D₁] (99% D) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoroacetic [2H]acid, TFA-D, Trifluoroacetic acid-[²H₁]

16640-0100 166400050 166400250 166401000
TS16640-0100EA 54 USD
TS16640-0100 TS166400050 TS166400250 TS166401000
Trifluoroacetic acid-[D₁] (99% D) for NMR spectroscopy
Trifluoroacetic acid-[D₁]
Formula: F₃CCOOD
MW: 115.02 g/mol
Boiling Pt: 75
Melting Pt: -15
Density: 1.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044563
CAS Number: 599-00-8
EINECS: 209-961-2
UN: 2699
ADR: 8 I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR