Acetic acid-[D1] (98% D) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

16620-0500 166202500
TS16620-0500EA 92 USD
TS16620-0500 TS166202500
Acetic acid-[D1] (98% D) for NMR spectroscopy
Acetic acid-[D1]
Formula: C₂H₃DO₂
Boiling Pt: 116…117 °C (760 mmHg)
Melting Pt: 15…16 °C
Density: 1.07 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00051050
CAS Number: 758-12-3
EINECS: 212-059-1
UN: 2789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤1 % (Coulometric)
D-Enrichment ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR