Dimethylamine 40% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

16367-0010 163670025
TS16367-0010EA 46 USD
TS16367-0010 TS163670025
Dimethylamine 40% in aqueous solution
Dimethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Melting Pt: –37 °C
Density: 0.89 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 39.0 to 41.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR