Diethylene glycol monobutyl ether 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

14971-0100 149710010 149710025
TS14971-0100EA 255 USD
TS14971-0100 TS149710010 TS149710025
Diethylene glycol monobutyl ether 99+%
Diethylene glycol monobutyl ether
Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 231 °C (760 mmHg)
Melting Pt: –68 °C
Density: 0.955 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 13,01556

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %
Color scale ≤10 APHA
Specific gravity (20 °C) 0.952 to 0.958
Acidity (CH3COOH) ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR