Thioacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13878-1000
138780025
138785000
TS13878-1000EA
54
USD
TS13878-1000
TS138780025
TS138785000
Thioacetic acid 98%
Thioacetic acid
Formula:
C₂H₄OS Boiling Pt: 87…93 °C (760 mmHg) Melting Pt: –17 °C Density: 1.07 g/cm³ Flash Pt: 18 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004853 CAS Number: 507-09-5 EINECS: 208-063-8 UN: 2436 ADR: 3,II Merck Index: 12,09454 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4650 (20 °C, 589 nm) |
Titration Argentometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...