Styrene 99% stabilized, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylethene, Phenylethylene, Vinylbenzene

13279-0010 13279-0100 132790025 132790250
TS13279-0010EA 58 USD
TS13279-0010 TS13279-0100 TS132790025 TS132790250
Styrene 99% stabilized, Extra Pure
Styrene
Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 145…146 °C (760 mmHg)
Melting Pt: –31 °C
Density: 0.906 g/cm³
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008612
CAS Number: 100-42-5
EINECS: 202-851-5
UN: 2055
ADR: 3,III
Merck Index: 13,08944

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5480 (20 °C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA
Stabilizer 10 to 20 ppm p-tert-Butylcatechol
Peroxides ≤0.01 % (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR