2'-Methoxyacetophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12567-0250
125671000
TS12567-0250EA
76
USD
TS12567-0250
TS125671000
2'-Methoxyacetophenone 99%
2'-Methoxyacetophenone
Formula:
C₉H₁₀O₂ Boiling Pt: 131 °C (18 mmHg) Density: 1.09 g/cm³ Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008725 CAS Number: 579-74-8 EINECS: 209-446-2 |
Specification Test Results
Appearance | Clear light yellow to orange to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5383 to 1.5403 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...