Ethylbenzene 99.8%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylethane

11808-5000 118080025 118080250 118080251
TS11808-5000EA 60 USD
TS11808-5000 TS118080025 TS118080250 TS118080251
Ethylbenzene 99.8%, pure
Ethylbenzene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.86 g/cm³
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3,II
Merck Index: 13,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4970 (20 °C, 589 nm)
Water ≤0.02 % (Coulometric)
GC ≥99.75 %
Color scale ≤10 APHA
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Silicon dioxide (SiO2) ≤50 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.01 %
Loss on heating ≤1.0 % (285 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR