1,2-Dichloroethane 99.8+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

11336-1000 113360010 113360025
TS11336-1000EA 73 USD
TS11336-1000 TS113360010 TS113360025 TS113360250
1,2-Dichloroethane 99.8+%, Extra Pure
1,2-Dichloroethane
Formula: ClH₂CCH₂Cl
Boiling Pt: 81…85 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.25 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3,II
Merck Index: 13,03831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48789e88e6041156e33712fd7c0198d7a4910266&vid=4238623c54017660b83d3baa856a69cc1aa8ee6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20 °C, 589 nm)
Water ≤0.02 % (Coulometric)
GC ≥99.80 %
Color scale ≤25 APHA
Residue after evaporation ≤0.005 %
Free acid (HCl) ≤0.01 %
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR