Benzonitrile 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

10554-0010 105540025
TS10554-0010EA 80 USD
TS10554-0010 TS105540025
Benzonitrile 99%, pure
Benzonitrile
Formula: C₇H₅N
Boiling Pt: 188…191 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1.01 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001770
CAS Number: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: 6.1,II
Merck Index: 13,01098

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5290 (20 °C, 589 nm)
GC ≥98.5 %
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR