Potassium acetate, Molecular Biology Grade, Calbiochem®, Millipore®
Supplier: MilliporeSigma
Synonyms:
Acetic acid potassium salt
529543-1KG
529543-250GM
80503-238EA
215.34
USD
80503-238
80503-240
Potassium acetate, Molecular Biology Grade, Calbiochem®, Millipore®
Potassium acetate
Formula:
CH₃COOK MW: 98.14 g/mol |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...