1-(3-sulfopropyl)pyridinium hydroxide inner salt

Supplier: MilliporeSigma

Synonyms: NDSB 201, 3-(1-Pyridinio)propanesulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulphonate

480005-250GM 480005-25GM
80058-456EA 500.72 USD
80058-456 80058-458
1-(3-sulfopropyl)pyridinium hydroxide inner salt
1-(3-sulfopropyl)pyridinium hydroxide inner salt
Formula: C₈H₁₁NO₃S
MW: 201.24 g/mol
MDL Number: MFCD00064468
CAS Number: 15471-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR