(2-Nitrophenyl)-β-D-galactopyranoside (ONPG), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

M-Clarity™ quality level = MQ100

48712-1GM 48712-5GM
80057-142EA 56.76 USD
80057-142 80057-144
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG), Calbiochem®, Sigma-Aldrich®
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)

A β-galactosidase substrate for colorimetric and EIA applications; counterpart of widely used pNPP/alkaline phosphatase substrate. o-Nitrophenol is produced as the end product and is monitored at 405 nm.


Contaminants - free o-nitrophenol: ≤0.05%

Formula: C₁₂H₁₅NO₈
Storage Temperature: Refrigerator
MDL Number: MFCD00063255
CAS Number: 369-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR