(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulfonate)
Supplier: MilliporeSigma
Synonyms:
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropanesulphonate), CHAPSO
220202-5GM
220202-1GM
80056-162EA
613.55
USD
80056-162
80056-160
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulfonate)
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Formula:
C₃₂H₅₈N₂O₈S MW: 630.89 g/mol |
CAS Number:
82473-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...