Creatine phosphate disodium salt, Millipore®

Supplier: MilliporeSigma

Synonyms: Phosphocreatine disodium hydrate, Sodium 2-(1-methyl-3-phosphonatoguanidino)acetate hydrate(2:1:x), N-(Imino[phosphonoamino]methyl)-N-methylglycine hydrate, Phosphocreatine disodium salt hydrate

2380-5GM 2380-25GM
80051-260EA 130 USD
80051-260 80051-258
Creatine phosphate disodium salt, Millipore®
Creatine phosphate disodium salt hydrate
Formula: C₄H₈N₃Na₂O₅P·H₂O
MW: 273.09 g/mol
MDL Number: MFCD00150191
CAS Number: 19333-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR