Wisp™ Style, Amber and Clear Vials, Chemglass
Supplier: Chemglass
CV-1256-1545 CV-1258-1545 CV-1259-1545
89493-008PK 38.04 USD
89493-008 75820-638 75820-640
Wisp™ Style, Amber and Clear Vials, Chemglass
Vials
This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers.
  • Clear or amber borosilicate
  • Available with graduated marking spots
These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.