AlphaTec® CSM Isolator Gloves, 85-304 Series, Ansell
Supplier: Ansell Healthcare
85304095 85304110
76408-476CS 6935.2 USD
76408-476 76408-478
AlphaTec® CSM Isolator Gloves, 85-304 Series, Ansell
Gloves
Non-clean/non sterile CSM RABS/Isolator gloves for use in 305 mm (12") ports.
  • Designed for high resistance against concentrated acids and bases
  • Port size: 305 mm (12")
  • Soft, flexible CSM material designed for ease of use
  • Resists ageing from exposure to oxygen, UV rays, and ozone
  • White colouring for easy detection of contamination
AlphaTec® CSM RABS and Isolator gloves are designed for high resistance against concentrated acids and bases. They are made from soft, flexible material designed for ease of use.
Certificaciones: CE 0493, EN 374:2003, EN ISO 374-5:2016, EN 388:2003, EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, Category III, UKCA 0321.
Envase: 1 pair per black sealed plastic bag, 10 pairs per shipper carton.