Copper disodium EDTA Cu 14.5%-15.5%

Supplier: Ambeed
Warning

A1000022-100G A1000022-25G A1000022-500G
76645-824EA 9 USD
76645-824 76645-826 76645-828
Copper disodium EDTA Cu 14.5%-15.5%
Copper disodium EDTA

Precaución: for Research Use Only

Formula: C₁₀H₁₂CuN₂Na₂O₈
MW: 397.7364 g/mol
MDL Number: MFCD00148920
CAS Number: 14025-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR