QUICKWIK™ Universal Granular Absorbent 20 lbs. Bags, Brady Worldwide
Supplier: Brady Worldwide
QW-20 QW-20H
76637-568PL 1559.88 USD
76637-568 76637-580
QUICKWIK™ Universal Granular Absorbent 20 lbs. Bags, Brady Worldwide
Spill Control Materials
Take care of industrial spills, leaks, and drips with QUICKWIK™ Universal Granular Absorbent bags, which offer more than 4X the absorption of clay granular and produces no carcinogens or dust.
  • Premium absorbent made of 100% recycled material
  • Certified safe for landfill disposal
  • Lightweight and easy to deploy
Effectively absorbs hydrocarbons, water-based fluids, and most aggressive chemicals.
Información de suministro: Sold and shipped as a pallet.