Racks for Freezers
Supplier: Eppendorf
6001000310 6001000311 6001000410 6001000411 6001072310 6001072410 6001072510 6001072910 6001081110 6001081210 6001081310 6001081410 6001081510 6001081910 6001000911 6001011110 6001011210 6001011310 6001011410 6001011510 6001011910 6001012210 6001012310 6001012410 6001022910 6001050010 6001050011 6001050110 6001050111 6001050210 6001050211 6001082210 6001082310 6001082410 6001082510 6001082910 6001012510 6001012910 6001021110 6001021210 6001021310 6001021410 6001021910 6001022210 6001022310 6001022410 6001071110 6001071210 6001071310 6001071410 6001071510 6001071910 6001072210 6001000510 6001000910
76545-676EA 124.99 USD
76545-676 76545-678 76545-680 76545-682 76545-788 76545-790 76545-792 76545-794 76545-796 76545-798 76545-800 76545-802 76545-804 76545-806 76545-688 76545-690 76545-692 76545-694 76545-696 76545-698 76545-700 76545-702 76545-704 76545-706 76545-728 76545-742 76545-744 76545-746 76545-748 76545-750 76545-752 76545-808 76545-810 76545-812 76545-814 76545-816 76545-708 76545-710 76545-712 76545-714 76545-716 76545-718 76545-720 76545-722 76545-724 76545-726 76545-774 76545-776 76545-778 76545-780 76545-782 76545-784 76545-786 76545-684 76545-686
Racks for Freezers
Racks Cryogenic Box Racks
These racks are heavy-duty and designed for higher capacity and durability. Specially designed for optimised cooling conditions and improved sample safety.
  • Fit freezer boxes up to a 136×136 mm for increased storage flexibility
  • Drawer-style freezer racks, each drawer can be individually removed, leaving the rest in the freezer
  • In side-access racks each box fits into a distinct, individual compartment for easier inventory tracking
There are two systems used in laboratories - the standard 4-boxes deep racks and 5-boxes deep racks. The easiest way is to take out all racks (with the samples/ boxes included) and store those racks in some backup freezer. To be compatible with most common ULT freezer suppliers, these ULT freezers are designed for 4-boxes deep racks.
The ULT freezer with 3-compartments/shelves has higher compartments compared to the 5-compartment ULT freezer. Racks of a 5-compartment freezer do fit into a 3-compartment freezer but not vice versa. Most of the storage boxes have to be pulled out of the in the cold inner area of the ULT freezer to access the samples.
To get the maximum capacity out of the ULT freezer, the CryoCube F740 series (3-compartment) can be equipped with 3-compartment MAX racks. These racks only fit into the CryoCube F740 series 3-compartment instruments.
Información de suministro: Locking rods are now available for chest freezer racks to provide additional sample security.