Gas Permeable Heat Sealing Membranes, Pierceable, Peelable

Supplier: AZENTA US, INC
4TI-0597 4TI-0597/ST 4TI-0598 4TI-0598/122 4TI-0598/S 4TI-0598-SLR 4TI-0599 4TI-0599/S 4TI-0599-SLR
76637-910CS 127.08 USD
76637-910 76637-808 76637-954 76637-758 76637-904 76637-762 76637-792 76637-782 76637-776
Gas Permeable Heat Sealing Membranes, Pierceable, Peelable
Seals Microplate Seals
The gas permeable heat seal is made from a woven material and is designed for use in cell culture, due to its porous nature. Suitable for cell culture, overnight incubations, as well as insect and seed storage.

  • Compatible with polypropylene, polystyrene and cyclic olefin copolymer (COC) plates
  • Pierceable with a pipette tip, manually or by liquid handling robots
  • Peelable
  • Autoclavable (121 °C)
  • Gas permeability rate: 180 m³/m²/day
  • Moisture vapor transmission rate: 20 g/m²/day
  • Effective for overnight incubations
  • Temperature range: –20 to +80 °C

Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective.

The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.

Información para pedidos: Available as sheets for use with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR