Heat Sealing Foils, Pierceable, Peelable
Supplier: AZENTA US, INC
4TI-0535 4TI-0535/122 4TI-0535/S 4TI-0535-SLR 4TI-0536 4TI-0537 4TI-0537/S
76637-922EA 1266.99 USD
76637-922 76637-846 76637-918 76637-712 76637-778 76637-928 76637-916
Heat Sealing Foils, Pierceable, Peelable
Seals Microplate Seals
The aluminum heat sealing foil is resealable, peelable, pierceable, suitable for compound storage and PCR. Suitable for short term storage.
  • Pierceable with a pipette tip, manually or by liquid handling robots
  • Peelable
  • Seals polypropylene and polystyrene plates
  • Resealable by applying another Seal straight on top of a previously pierced seal
  • Moderate solvent resistance
  • Autoclavable (121 °C)
  • Red stripe on the sheet foils clearly indicates the non-sealing surface, for ease of seal orientation and handling
  • Temperature range: –20 to +110 °C
  • Non sterile
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective.
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
Información para pedidos: Available as sheets for use with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers. Gamma treatment available upon request.