Heat Sealing Foils, Pierceable
Supplier: AZENTA US, INC
4TI-0530 4TI-0530/122 4TI-0530/S 4TI-0531 4TI-0531/GR 4TI-0531/RA 4TI-0532 4TI-0532/S
76637-822EA 1266.99 USD
76637-822 76637-940 76637-944 76637-764 76637-866 76637-736 76637-832 76637-876
Heat Sealing Foils, Pierceable
Seals Microplate Seals
The pierceable foil heat seal is compatible with polypropylene and polystyrene plates.
  • Pierceable with a pipette tip manually or by a liquid handling robot
  • Resealable by applying another pierce seal straight on top of a previously pierced seal
  • Autoclavable (121 °C)
  • Blue stripe on the sheet foils clearly indicates the non-sealing surface for ease of seal orientation and handling
  • Available with a printed grid reference on the non-sealing surface
  • Temperature range: –20 to +120 °C
  • Non sterile
Heat sealing offers a 100% effective method of plate sealing,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable