Heat Sealing Films, Clear
Supplier: AZENTA US, INC
4TI-0540 4TI-0540/80 4TI-0540/S 4TI-0541 4TI-0542 4TI-0542/S
76637-788EA 1284.91 USD
76637-788 76637-936 76637-930 76637-912 76637-836 76637-828
Heat Sealing Films, Clear
Seals Microplate Seals
The clear heat seal is an optically clear laminate film forms a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) SBS format plates.
  • Samples can be accessed by pre-piercing with a blade, needle or the pierce plate (4ti-0398)
  • Moderate solvent resistance and can be utilized for short term compound storage at room temperature
  • Temperature range: –80 to +80 °C
  • Non sterile
Optical clarity enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays.
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. These clear heat seals form a complete seal to a plate enabling both low temperature uses, including low temperature storage, and high temperature uses, such as PCR (when used with a pressurized heated lid).
The sheet seals fit all standard SBS footprint plates from 12 to 1536 wells and are inter-leaved with paper sheets to help denote which side is the sealing side and aid removal of one sheet at a time from the pack.
Información para pedidos: Available as sheets for use with manual and semiautomated sealers such as the semi-automated sheet heat sealer. Also available in multiple roll formats compatible with specified automated heat sealers such as the automated roll heat sealer. Gamma treatment available upon request.