Anti-Cytokeratin, Pan Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems

RP167-01 RP167-05 RP167
76635-168EA 108.26 USD
76635-168 76635-170 76635-172
Anti-Cytokeratin, Pan Rabbit Polyclonal Antibody
Antibodies
This antibody recognizes cytokeratin polypeptides ranging from 40 through 58 kDa, corresponding to cytokeratins 5, 6, 8,17 and 19. It shows a broad pattern of reactivity with human epithelial tissues from simple glandular epithelia to stratified squamous epithelia, like epidermis, mammary gland ducts and tracheal epithelium.
Order Now

SPECIFICATIONS

Antigen Name Cytokeratin, Pan
Environmentally Preferable
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR