Anti-P40 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP163 RP163-01 RP163-05
76635-166EA 750.15 USD
76635-166 76635-162 76635-164
Anti-P40 Rabbit Polyclonal Antibody
Antibodies
p40 is a relatively unknown antibody that recognizes ΔNp63-a p63 isoform suggested to be highly specific for squamous/basal cells. In a recent study, p40 is equivalent to p63 in sensitivity for squamous cell carcinoma, but it is markedly superior to p63 in specificity1, which eliminates a potential pitfall of misinterpreting a p63-positive adenocarcinoma or unsuspected lymphoma as squamous cell carcinoma. These findings strongly support the routine use of p40 in place of p63 for the diagnosis of pulmonary squamous cell carcinoma.