Anti-S-100 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP035-05 RP035
76635-024EA 85 USD
76635-024 76635-026 76635-028
Anti-S-100 Rabbit Polyclonal Antibody
Antibodies
This antibody is specific against an epitope located on the β chain (i.e. in S-100a and S-100b) but not on the α chain of S-100 (i.e. in S-100a and S-100ao). This antibody can be used to localize S-100a and S-100b in various tissue sections.