Powerbanket IBC tote warmers offer cutting-edge technology for freeze protection, chemical storage, and viscosity control in a flexible, easy-to-install design that will effectively, evenly, and reliably heat your IBC tote. Our patented design eliminates hot and cold spots by applying uniform full-coverage heat across the entire surface of the container, allowing users to keep their materials at the proper viscosity while remaining at specified thermal levels. Each tote heater can be equipped with either an adjustable or an IoT smart thermostat. Tote heaters come in two voltages: 120 volts and 240 volts.
- Powerblanket - designed for any definitive heating applications, surrounded by a rugged vinyl shell in a wraparound design with a removable top, safely raises material temperatures as high as 145 ºF and can be used in temperatures as low as −20 ºF
- Powerblanket Xtreme - designed for harsh climates, surrounded by a rugged vinyl shell in a wraparound design with a removable top, safely raises material temperatures as high as 145 ºF and can be used in temperatures as low as −20 ºF
- Durable and weather-resistant
- Safely heat and protect a wide variety of chemicals and materials
- Includes adjustable digital thermostatic controller
- Insulated full-wrap design
- Easy access with a removable top
- Deliver uniform heat to temperature-sensitive products
- Preserve expensive materials without scorching or burning
- Prevent waste by maintaining consistent temperatures
- Raise material temperatures as high as 145 ºF / 62 ºC (± 5 ºF/3 ºC), depending on ambient conditions
Certificaciones: ETL certified to UL and CSA safety standards.