Anti-Cytokeratin 7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]
Supplier: Diagnostic Biosystems
MOB057-01 MOB057-05 MOB057
76633-176EA 108.26 USD
76633-176 76633-178 76633-180
Anti-Cytokeratin 7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]
Antibodies
This antibody recognizes keratin polypeptide of 54 kDa. It stains a number of epithelial cells including ductal and glandular epithelia.