Powerblanket® Lite Pail Heaters
Supplier: POWERBLANKET LLC MS
PBL2G PBL1G
104018-926EA 170.15 USD
104018-926 104018-954
Powerblanket® Lite Pail Heaters
Heaters Heating Mantles
Powerblanket’s bucket heaters provide a range of heating solutions for warming and storing temperature-sensitive materials. Our patented design eliminates hot and cold spots by applying uniform full-coverage heat across the entire surface of the container, allowing users to keep their materials at the proper viscosity while remaining at specified thermal levels. Each drum heater can be equipped with either an adjustable or a fixed thermostat.
  • Powerblanket Lite - designed for light residential and industrial applications, low power density.
  • Powerblanket - designed for any definitive heating applications, surrounded by a rugged vinyl shell, safe to use in temperatures as low as −20 ºF
  • Powerblanket Xtreme - designed for harsh climates, safe to use in temperatures as low as −40 Fº
  • Adjustable temperature from ambient to 145 ºF on PRO models
  • Are safe to use on both steel and poly buckets
  • Provide an insulated full-wrap design making it more efficient
  • Deliver uniform heat to temperature-sensitive products
  • Preserve expensive materials without scorching or burning
  • Prevent product waste by maintaining consistent temperatures
  • Cinch straps to secure tight fit
  • Safe for outdoor use
  • Water-resistant
  • Hassle free, preset internal 100 °F temperature on RR Rapid Ramp models
  • Hassle free, preset internal 145 °F temperature on PBL05 insulated band heater model
Bucket heaters come in two voltages: 120 and 240 V.
All the bucket heaters come as an insulated full-wrap design with cinch straps to secure a tight fit. They're also water-resistant and can be used on poly and steel buckets.
Certificaciones: All Powerblanket products are ETL certified to UL, CSA, and CE safety standards.