PCR Plates, 384-Well, Fully Skirted, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-1384 4TI-1385 4TI-1387
76638-096CS 253.67 USD
76638-096 76637-976 76637-986
PCR Plates, 384-Well, Fully Skirted, Azenta Life Sciences
Microplates PCR Plates
These plates are designed for use on standard 384-well thermal cyclers. Polypropylene, cut corner A24; working volume: <30 µl, total well capacity: 55 µl.
  • Subject to a strict QC procedure which includes a visual check and leak testing of every tube
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • 30 µl working volume, 55 µl total well capacity when used with sealing options
  • Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
  • Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
  • Compatible with blocks designed for 384-well PCR plates and standard multichannel pipettes
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility and as such comply with the same stringent requirements as the FrameStar® range.
Información para pedidos: Available barcoded upon request.