FrameStar® PCR Plates, 96-Well, Semi-Skirted, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0900/C 4TI-0901
76629-076CS 307.46 USD
76629-076 76629-090
FrameStar® PCR Plates, 96-Well, Semi-Skirted, Azenta Life Sciences
Microplates PCR Plates
96-well universal semi-skirted plate designed for use on standard thermal cyclers, cut corner H1. Ideal for use with robotic systems. Compatible with standard multichannel pipettes.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Stackable
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • <0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample evaporation for improved consistency in PCR results
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
The rigid FrameStar two-component design eliminates warping and distortion during PCR, making it ideal for use with robotic systems.
The semi-skirted allows for labeling or barcoding for sample tracking.
Información para pedidos: Available barcoded upon request.