8-Well PCR Tube Strips with Attached Caps, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0792 4TI-0793 4TI-0794
76638-064CS 147.96 USD
76638-064 76638-124 76638-156
8-Well PCR Tube Strips with Attached Caps, Azenta Life Sciences
Tubes PCR Tubes
Clear polypropylene tube strips with tethered domed or optically clear flat caps.
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Tubes on each strip are joined together either by three links (flat cap variety - 4ti-0792 and 4ti-0793) or by one strong link (domed cap variety - 4ti-0794) to make the strip more rigid and help reduce any chance of spillage
  • Each cap is separately joined to a tube, making it impossible to either cross contaminate another tube with the wrong cap, or to lose a cap altogether
  • The tube strips can be easily separated by cutting the links, to make smaller sections or individual tubes
  • The flat optically clear caps enable light signals, such as fluorescence, to pass through without affecting the signal, and are suitable for imaging techniques including RT-qPCR
These PCR tubes are molded from virgin polypropylene in a UK-based Class 7 ISO certified cleanroom production facility, and comply to the same stringent requirements as the FrameStar range.
Recommended for low to medium throughput applications.