PCR Plates, 96-Well, Skirted, Low Profile, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0740 4TI-0741
76638-132CS 233.4 USD
76638-132 76638-140
PCR Plates, 96-Well, Skirted, Low Profile, Azenta Life Sciences
Microplates PCR Plates
Universal 96-well skirted plate designed for use on standard thermal cyclers. 0.1 ml wells, polypropylene, cut corner H1; working volume: <100 µl, total well capacity: 200 µl.
  • Subject to a strict QC procedure which includes a visual check and leak testing of every tube
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Low profile 0.1 ml (100 µl) working volume, 0.2 ml (200 µl) total well capacity when used with sealing options
  • Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
  • Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
  • Compatible with blocks designed for skirted PCR plates and standard multichannel pipettes
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility, and as such comply with the same stringent requirements as the FrameStar® range.
Información para pedidos: Available barcoded upon request.