Formical-2000™ Decalcifier, StatLab
Supplier: StatLab
A fast yet gentle bone decalcifier that has been formulated from semiconductor grade formic acid, highly purified reagent grade water and EDTA. Formical-2000™ works well not only as a general purpose decalcifier for all routine histological specimens, it works especially well to decalcify tissue for immunohistochemistry. Will safely decalcify bone without destroying antigen sites.
Envase: Manufactured in the USA