Sample Storage Cap Carriers, SBS, Azenta Life Sciences
Supplier: AZENTA US, INC
65-9801 65-9802 68-53111-10N 68-53111-10X 68-53111-50N 68-53111-50X 67-63111-10 67-63111-50 66-9951
76636-820CS 654.94 USD
76636-820 76636-908 76636-900 76636-586 76636-958 76636-822 76636-858 76636-954 76636-770
Sample Storage Cap Carriers, SBS, Azenta Life Sciences
Crimpers, Cappers and Decappers
Improve process effeciency. SBS-format screw cap carriers enable automated capping of 96-format sample storage tubes.
  • Cap Carriers enable uncapped tubes to be used immediately and then capped using an automated capping and de-capping system
  • Leads to improved process efficiency and allows batch filling of tubes prior to capping
  • When used manually, by transferring caps from the carrier to the tube using a single tube transfer device, the operation becomes faster and the risk of contamination is reduced as cap handling is eliminated
Cap carriers are suitable for manual use or for use with semi-automated screw cap decapper or recapper, single channel systems and automated single column screw cap capper or decapper systems.