Decal™ Decalcifier, StatLab
Supplier: StatLab
1211-1
1211-32
1211-5
76547-992EA
76.91
USD
76547-992
76547-994
76547-996
Decal™ Decalcifier, StatLab
Stains
Microscopy and staining reagents
Since 1957 Original Decal® has been the industry standard. A gentle decalcifier formulated from semiconductor grade hydrochloric acid, EDTA, and highly purified reagent grade water. Can be used to safely decalcify all types of tissue samples from needle biopsies to cortical bone. It’s mild enough that most samples can be left in overnight; over decalcification is not a worry.
Envase: Manufactured in the USA