Essentials Disposable Micro Spatulas, Sterile, PS, Antylia Scientific
Supplier: Antyila Scientific
06264-20 06264-21 06264-22 06264-23
76533-660PK 181.04 USD
76533-660 76533-662 76533-664 76533-666
Essentials Disposable Micro Spatulas, Sterile, PS, Antylia Scientific
Spatulas
Dispense and sample small quantities of powders and granules.
  • Manufactured and packed in a Class 100,000 cleanroom to ensure cleanliness and reduce cross contamination
  • Made from high impact polystyrene (HIPS) which conforms to FDA, EC, and EU regulations
  • BSE/TSE free
The single-use spatulas are gamma irradiated and individually packaged so they can be used straight out of the bag without cleaning.
Certificaciones: Supplier Certificate of Conformity is included with product shipment.