Z-Tle-OH (dicyclohexylammonium) salt ≥99.0% (by HPLC), Sigma-Aldrich®

Synonyms: Z-tBu-Gly-OH·DCHA, Z-Tle-OH·DCHA, (-)-N-Benzyloxycarbonyl-L-tert-leucine dicyclohexyl ammonium salt, Dicyclohexylamine (S)-2-(((benzyloxy)carbonyl)amino)-3,3-dimethylbutanoate, Z-L-tert-Leucine (dicyclohexylammonium) salt

M-Clarity™ quality level = MQ100

96720-25G 96720-5G
220101-068EA 1550.77 USD
220101-068 220101-069
Z-Tle-OH (dicyclohexylammonium) salt ≥99.0% (by HPLC), Sigma-Aldrich®
Z-Tle-OH (dicyclohexylammonium) salt
Formula: C₁₄H₁₉NO₄·C₁₂H₂₃N
MW: 446.632 g/mol
Melting Pt: 163-167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00161480
CAS Number: 62965-37-1

Order Now

Specification Test Results

Solubility Soluble (20 °C)
Melting Point 164 - 167 °C
Molar Mass 446.62 g/mol
pH 6,3 (H2O, 20 °C) (saturated solution)
Melting point +164...+167 °C
Assay (HPLC, area%) ≥98% (a/a)
Specific rotation -7 to -6°
Identity (IR) Passes test
Storage temperature +15...+25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR