2,4-Difluoro-5-maleimidobenzoic acid ≥95%

Supplier: AFG BIOSCIENCE LLC

C59222-500MG C59222-1G C59222-2G
76620-350EA 550.84 USD
76620-350 76620-352 76620-354
2,4-Difluoro-5-maleimidobenzoic acid ≥95%
2,4-Difluoro-5-maleimidobenzoic acid
Formula: C₁₁H₅F₂NO₄
MW: 253.16 g/mol
Storage Temperature: Refrigerator
CAS Number: 1178844-49-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR