NH2-Gly-Gly-PEG3-Azide ≥95%

Supplier: AFG Bioscience

C56985-100MG C56985-25MG
76620-320EA 591.51 USD
76620-320 76620-318
NH2-Gly-Gly-PEG3-Azide ≥95%
NH2-Gly-Gly-PEG3-Azide
Formula: C₁₂H₂₄N₆O₅
Storage Temperature: Refrigerator
CAS Number: 1945990-18-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Colorless Sticky Oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR