t-​Boc-​N-​amido-​PEG1-​Azide ≥95%

Supplier: AFG BIOSCIENCE LLC

C56952-1G C56952-500MG
76620-316EA 542.22 USD
76620-316 76620-314
t-​Boc-​N-​amido-​PEG1-​Azide ≥95%
t-​Boc-​N-​amido-​PEG1-​Azide
Formula: C₉H₁₈N₄O₃
MW: 230.26 g/mol
Storage Temperature: Refrigerator
CAS Number: 176220-30-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Pale Yellow Oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR