2-​Azidoethan-​1-​amine 95%

Supplier: AFG Bioscience

C56668-1G C56668-2G
76620-274EA 400.61 USD
76620-274 76620-276
2-​Azidoethan-​1-​amine 95%
2-​Azidoethan-​1-​amine
Formula: C₂H₆N₄
Storage Temperature: Freezer
MDL Number: MFCD01727214
CAS Number: 87156-40-9

Order Now

Specification Test Results

Purity Limit 95%
Appearance yellow transparent liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR