3-​Deoxy-​D-​glycero-​D-​galacto-​2-​nonulosonic acid ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: KDN, Ketodeoxynonulosonic acid, Deaminoneuraminic acid

C53686-50MG C53686-100MG C53686-250MG
76619-950EA 250.38 USD
76619-950 76619-952 76619-954
3-​Deoxy-​D-​glycero-​D-​galacto-​2-​nonulosonic acid ≥95%
3-​Deoxy-​D-​glycero-​D-​galacto-​2-​nonulosonic acid
Formula: C₉H₁₆O₉
MW: 268.22 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01863086
CAS Number: 153666-19-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR