6-Fluoro-N-acetylgalactosamine Peracetate 95%

Supplier: AFG BIOSCIENCE LLC

C52506-10MG C52506-25MG
76619-806EA 550.84 USD
76619-806 76619-808
6-Fluoro-N-acetylgalactosamine Peracetate 95%
6-Fluoro-N-acetylgalactosamine Peracetate
Formula: C₁₄H₁₉FO₉
MW: 350.3 g/mol
Storage Temperature: Freezer
CAS Number: 63597-78-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR