Ac4GalNAl ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: N-(4-pentynoyl)-galactosamine tetraacetate

C52023-100MG C52023-50MG
76619-784EA 550.84 USD
76619-784 76619-782
Ac4GalNAl ≥95%
Ac4GalNAl
Formula: C₁₉H₂₅NO₁₀
MW: 427.4 g/mol
Storage Temperature: Freezer
CAS Number: 1658458-26-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR