Ac4GlcNAl ≥95%

Supplier: AFG Bioscience

Synonyms: Ac4GlcNAlk, (3R,4R,5S,6R)-6-(Acetoxymethyl)-3-(pent-4-ynamido)tetrahydro-2H-pyran-2,4,5-triyl triacetate, N-(4-pentynoyl)-glucosamine tetraacetate

C52022-100MG C52022-50MG C52022-25MG
76619-780EA 550.84 USD
76619-780 76619-778 77888-844
Ac4GlcNAl ≥95%
Ac4GlcNAl
Formula: C₁₉H₂₅NO₁₀
Storage Temperature: Freezer
MDL Number: MFCD32857621
CAS Number: 1361993-37-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Grey Amorphous Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR